EMAIL THIS PAGE TO A FRIEND
45790 Riedel-de Haën

1,3-Indandione

analytical standard, for environmental analysis

Synonym: 1,3-Dioxoindan

  • CAS Number 606-23-5

  • Empirical Formula (Hill Notation) C9H6O2

  • Molecular Weight 146.14

  •  Beilstein Registry Number 1210061

  •  EC Number 210-109-7

  •  MDL number MFCD00003779

  •  PubChem Substance ID 24869406

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

grade   analytical standard, for environmental analysis
mp   129-132 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?