CAS Number 21087-64-9
Beilstein Registry Number 746650
EC Number 200-835-2
MDL number MFCD00055525
| grade | PESTANAL®, analytical standard |
| concentration | 100 ng/μL in acetonitrile |
| storage temp. | 2-8°C |
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?