• USA Home
  • 46208 - Nitrofurazone solution

EMAIL THIS PAGE TO A FRIEND
46208 Riedel-de Haën

Nitrofurazone solution

100 ng/μL in acetonitrile, VETRANAL, analytical standard

Synonym: Nitrofural solution

  • CAS Number 59-87-0

  • Empirical Formula (Hill Notation) C6H6N4O4

  • Molecular Weight 198.14

  •  Beilstein Registry Number 1347298

  •  EC Number 200-835-2

  •  MDL number MFCD00003225

  •  PubChem Substance ID 24869937

Purchase

Properties

grade   VETRANAL, analytical standard
concentration   100 ng/μL in acetonitrile

Description

Legal Information

VETRANAL is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1648 3 / PGII
WGK Germany 
2
Flash Point(F) 
35.6 °F
Flash Point(C) 
2 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

31706 Nitrofurazone solution, 100 μg/mL in acetonitrile, VETRANAL, analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?