EMAIL THIS PAGE TO A FRIEND
E7023 Riedel-de Haën

Ethyl alcohol, Pure

200 proof, for molecular biology

Synonym: Ethanol, absolute alcohol, non-denatured ethanol

Purchase

Description

Frequently Asked Questions

Frequently Asked Questions are available for this Product.

Biochem/physiol Actions

Positive allosteric modulator of GABAA receptors, and negative allosteric modulator of NMDA glutamate receptors.

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Sales restrictions may apply

Legal Information

Riedel-de Haën is a registered trademark of Honeywell Specialty Chemicals Seelze GmbH.

Application

Pure ethyl alcohol (ethanol) has been used in the following process:
• DNA and RNA extraction
• array hybridization
• ethanol sedation assay

Suitable for use in the precipitation of nucleic acids

General description

This product is 200proof, non-denatured absolute ethanol (100%). This is a molecular biology grade product and is tested for the RNase and Dnase.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1170 - class 3 - PG 2 - Ethanol
WGK Germany 
1
RTECS 
KQ6300000
Flash Point(F) 
57.2 °F
Flash Point(C) 
14 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

also commonly purchased with this product

Product #

Description

Add to Cart

Z350370 Carestream® Kodak® BioMax® MR film, size 8 in. (20 cm) × 10 in. (25 cm)
59304 2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
I9516 2-Propanol, for molecular biology, BioReagent, ≥99.5%

recommended

Product #

Description

Add to Cart

CLS3401 Corning® Transwell® polycarbonate membrane cell culture inserts, 12 mm Transwell with 0.4 μm pore polycarbonate membrane insert, TC-treated, w/ lid, sterile, 48/cs

suggested gloves for splash protection

Product #

Description

Add to Cart

Z677353 Dermatril P nitrile gloves, size XS
Z677388 Dermatril P nitrile gloves, size M
Z677396 Dermatril P nitrile gloves, size L
Z677418 Dermatril P nitrile gloves, size XL

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?