EMAIL THIS PAGE TO A FRIEND
00138 Sigma-Aldrich

Dihydrojasmone

analytical standard

Synonym: 3-Methyl-2-pentyl-2-cyclopenten-1-one

  • CAS Number 1128-08-1

  • Linear Formula CH3(CH2)4C5H4(CH3)(=O)

  • Molecular Weight 166.26

  •  Beilstein Registry Number 1906471

  •  EC Number 214-434-5

  •  MDL number MFCD00036480

  •  PubChem Substance ID 24844974

Purchase

Properties

Related Categories A-F, A-L, Additional Standards, All Alphabetically Sorted, Analytical Standards,
grade   analytical standard
InChI Key   YCIXWYOBMVNGTB-UHFFFAOYSA-N
assay   ≥98.0% (GC)
shelf life   limited shelf life, expiry date on the label
refractive index   n20/D 1.479(lit.)
  n20/D 1.481
bp   120-121 °C/12 mmHg(lit.)
density   0.916 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
GY7302000
Flash Point(F) 
204.8 °F
Flash Point(C) 
96 °C
Protocols & Articles

Related Content

Flavor and Fragrance Analysis

It’s all about your workflow. Sample preparation, separation, detection and analysis. Speed and resolution is key in determining purity or verifying composition.
Keywords: Calorimetry, Gas chromatography, Mass spectrometry, Sample preparations, Separation

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?