• USA Home
  • 00188 - 3′-Acetamidoacetophenone

EMAIL THIS PAGE TO A FRIEND
00188 Sigma-Aldrich

3′-Acetamidoacetophenone

≥98.0% (GC)

Synonym: 3-Acetylacetanilide

  • CAS Number 7463-31-2

  • Empirical Formula (Hill Notation) C10H11NO2

  • Molecular Weight 177.20

  •  Beilstein Registry Number 2363443

  •  MDL number MFCD00032278

  •  PubChem Substance ID 329747013

Purchase

Properties

Related Categories Amides, Building Blocks, C8 to C20+, Carbonyl Compounds, Chemical Synthesis,
InChI Key   AFZTYHRVDOKRKV-UHFFFAOYSA-N
assay   ≥98.0% (GC)
impurities   ≤0.5% water
mp   127-130 °C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?