• USA Home
  • 00213 - Benzoyl peroxide blend with dicyclohexyl phthalate

EMAIL THIS PAGE TO A FRIEND
00213 Sigma-Aldrich

Benzoyl peroxide blend with dicyclohexyl phthalate

technical, ~50% (T)

  • Empirical Formula (Hill Notation) C14H10O4

  • Molecular Weight 242.23

  •  MDL number MFCD00003071

  •  PubChem Substance ID 329747018

Purchase

Properties

Related Categories Chemical Synthesis, Oxidizing Agents, Peroxides, Synthetic Reagents
InChI Key   OMPJBNCRMGITSC-UHFFFAOYSA-N
grade   technical
assay   ~50% (T)
impurities   ≤0.5% water
storage temp.   2-8°C

Description

Packaging

in inert filler with plasticizer (dicyclohexyl phthalate)

Application

• Oxidizing agent
• Crosslinking agent
• Oxidizer in palladium-catalyzed oxidative allylic C-H silylation

Price and Availability


Laboratory Gloves

Pharma QC
Safety & Documentation

Safety Information

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?