• USA Home
  • 00225 - 3-Acetamidobenzoic acid

EMAIL THIS PAGE TO A FRIEND
00225 Sigma-Aldrich

3-Acetamidobenzoic acid

≥98.0% (HPLC)

  • CAS Number 587-48-4

  • Empirical Formula (Hill Notation) C9H9NO3

  • Molecular Weight 179.17

  •  Beilstein Registry Number 2210867

  •  EC Number 209-600-9

  •  MDL number MFCD00013983

  •  PubChem Substance ID 329747020

Purchase

Properties

Related Categories Aromatic Amino Acids, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   RGDPZMQZWZMONQ-UHFFFAOYSA-N
assay   ≥98.0% (HPLC)
mp   247-252 °C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?