EMAIL THIS PAGE TO A FRIEND
00231 Sigma-Aldrich

Fmoc-Ser(tBu)-OPfp

technical, ≥90% (HPLC)

Synonym: Fmoc-O-tert-butyl-L-serine pentafluorophenyl ester

  • CAS Number 105751-13-1

  • Empirical Formula (Hill Notation) C28H24F5NO5

  • Molecular Weight 549.49

  •  Beilstein Registry Number 3642202

  •  MDL number MFCD00062213

  •  PubChem Substance ID 329747021

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Serine More...
InChI Key   DOUJYVMLNKRFHE-IBGZPJMESA-N
grade   technical
assay   ≥90% (HPLC)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?