EMAIL THIS PAGE TO A FRIEND
00269 Sigma-Aldrich

Dextran

analytical standard, for GPC, 5,000

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Application CRMs, Biochemicals and Reagents,
grade   analytical standard
  for GPC
form   powder or crystals
CofA   exact values can be taken from the accompanying certificate of analysis
mol wt   Mn ~3,260
  Mp ~4,440
  Mw ~5,220
Mw/Mn   ~1.60

Description

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles

Articles

Dextran and Related Polysaccharides

Historically, dextrans had been long recognized as contaminants in sugar processing and other food production. The formation of dextran in wine was shown by Pasteur to be due to the activity of micro...
Vicki Caligur
BioFiles 2008, 3.10, 17.
Keywords: Adhesion, Applications, Chromatography, Clinical, Cosmetics, Derivatizations, Diffusion, Filtration, Immobilization, Industries, Ion Exchange, Ligands, Microscopy, Molecular biology, Molecular biology techniques, Nuclear magnetic resonance spectroscopy, Nucleic acid hybridization, Pharmaceutical, Precipitation, Purification, Separation, Size-exclusion chromatography, Spectroscopy, Sulfations, Type

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?