• USA Home
  • 00320 - S-Acetamidomethyl-L-cysteine hydrochloride

EMAIL THIS PAGE TO A FRIEND
00320 Sigma-Aldrich

S-Acetamidomethyl-L-cysteine hydrochloride

≥99.0% (AT)

Synonym: H-Cys(Acm).HCl

  • CAS Number 28798-28-9

  • Empirical Formula (Hill Notation) C6H12N2O3S · HCl

  • Molecular Weight 228.70

  •  Beilstein Registry Number 4015821

  •  EC Number 249-230-5

  •  MDL number MFCD00077080

  •  PubChem Substance ID 329747042

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Cysteine/Cystine, Peptide Chemistry More...
InChI Key   SZWPOAKLKGUXDD-JEDNCBNOSA-N
assay   ≥99.0% (AT)
optical activity   [α]20/D −28±2°, c = 1% in H2O
mp   ~165 °C (dec.)

Description

Other Notes

Protected cysteine; cleavage is performed with mercuric acetate or silver nitrate; Oxidative cleavage to disulfides

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?