EMAIL THIS PAGE TO A FRIEND
00350 Sigma-Aldrich

3-Acetamidophenol

technical, ≥98.0% (HPLC)

Synonym: 3′-Hydroxyacetanilide

  • CAS Number 621-42-1

  • Linear Formula CH3CONHC6H4OH

  • Molecular Weight 151.16

  •  Beilstein Registry Number 907998

  •  EC Number 210-687-0

  •  MDL number MFCD00002263

  •  PubChem Substance ID 329747047

Purchase

Properties

Related Categories Building Blocks, C6 to C8, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds,
InChI Key   QLNWXBAGRTUKKI-UHFFFAOYSA-N
grade   technical
assay   ≥98.0% (HPLC)
mp   145-148 °C(lit.)
  145-148 °C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
2
RTECS 
AE4100000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?