• USA Home
  • 00352 - Fmoc-β-styryl-Ala-OH

EMAIL THIS PAGE TO A FRIEND
00352 Sigma-Aldrich

Fmoc-β-styryl-Ala-OH

≥98.0% (HPLC)

Synonym: (S)-2-(Fmoc-amino)-5-phenyl-4-pentenoicacid, Fmoc-3-styryl-L-alanine

Purchase

Properties

Related Categories Alanine Derivatives, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   ZFMHHKMOLFNMMV-DDVUFSRBSA-N
assay   ≥98.0% (HPLC)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?