• USA Home
  • 00390 - α-Acetamidocinnamic acid

EMAIL THIS PAGE TO A FRIEND
00390 Sigma-Aldrich

α-Acetamidocinnamic acid

purum, ≥99.0% (T)

Synonym: N-Acetyldehydrophenylalanine

  • CAS Number 5469-45-4

  • Linear Formula C6H5CH=C(NHCOCH3)COOH

  • Molecular Weight 205.21

  •  Beilstein Registry Number 1912549

  •  EC Number 226-795-6

  •  MDL number MFCD00008680

  •  PubChem Substance ID 329747057

Purchase

Properties

Related Categories Building Blocks, C11 to C12, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   XODAOBAZOQSFDS-YFHOEESVSA-N
grade   purum
assay   ≥99.0% (T)
mp   188-190 °C(lit.)
  190-193 °C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?