• USA Home
  • 00471 - (R)-2-Fmoc-1,2,3,4-tetrahydronorharmane-3-carboxylic acid

EMAIL THIS PAGE TO A FRIEND
00471 Sigma-Aldrich

(R)-2-Fmoc-1,2,3,4-tetrahydronorharmane-3-carboxylic acid

>97.0%

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Other Unnatural Amino Acids & Derivatives, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   IHHULIKSMJSELI-RUZDIDTESA-N
assay   >97.0%
optical activity   [α]20/D −56±2°, c = 1% in DMF
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?