• USA Home
  • 00535 - 6-Methyl-1,3,5-triazine-2,4-diamine

EMAIL THIS PAGE TO A FRIEND
00535 Sigma-Aldrich

6-Methyl-1,3,5-triazine-2,4-diamine

purum, ≥99.0% (NT)

Synonym: 2,4-Diamino-6-methyl-1,3,5-triazine, Acetoguanamine

  • CAS Number 542-02-9

  • Empirical Formula (Hill Notation) C4H7N5

  • Molecular Weight 125.13

  •  Beilstein Registry Number 118348

  •  EC Number 208-796-3

  •  MDL number MFCD00023192

  •  PubChem Substance ID 329747090

Purchase

Properties

InChI Key   NJYZCEFQAIUHSD-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (NT)
mp   274-276 °C(lit.)
  274-276 °C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?