EMAIL THIS PAGE TO A FRIEND
00628 Sigma-Aldrich

Iron

wire, purum p.a.

Purchase

Properties

Related Categories 26: Fe, Iron, Magnetic Materials, Magnetic Precursors, Materials Science,
InChI Key   XEEYBQQBJWHFJM-UHFFFAOYSA-N
grade   purum p.a.
form   wire
resistivity   9.71 μΩ-cm
diam.   0.2 mm
bp   2750 °C(lit.)
mp   1535 °C(lit.)
density   7.86 g/mL at 25 °C(lit.)
cation traces   Cu: ≤1000 mg/kg
  Mn: ≤7000 mg/kg
  Ni: ≤1000 mg/kg

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.




Ultra-High Purity Inorganics
Safety & Documentation

Safety Information

WGK Germany 
nwg
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?