EMAIL THIS PAGE TO A FRIEND
00632 Sigma-Aldrich

Iron

purum p.a., ≥98.0% (KT)

Purchase

Properties

InChI Key   XEEYBQQBJWHFJM-UHFFFAOYSA-N
grade   purum p.a.
assay   ≥98.0% (KT)
resistivity   9.71 μΩ-cm
diam.   0.57 mm
bp   2750 °C(lit.)
mp   1535 °C(lit.)
density   7.86 g/mL at 25 °C(lit.)
cation traces   Cu: ≤1000 mg/kg
  Mn: ≤7000 mg/kg
  Ni: ≤1000 mg/kg

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

356832 Iron, wire, diam. 0.5 mm, ≥99.9% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?