• USA Home
  • 00648 - Potassium hydroxide solution

EMAIL THIS PAGE TO A FRIEND
00648 Sigma-Aldrich

Potassium hydroxide solution

28.5%, for flue-gas analysis

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Application Specific Reagents, Decomposition Reagents
InChI Key   KWYUFKZDYYNOTN-UHFFFAOYSA-M
assay   28.5%
  28.5%±0.5% (T)

Description

Application

for flue-gas analysis

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1814 8 / PGII
WGK Germany 
1
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?