EMAIL THIS PAGE TO A FRIEND
00652 Sigma-Aldrich

Iodoacetic acid

puriss. p.a., ≥99.5% (T)

  • CAS Number 64-69-7

  • Linear Formula ICH2CO2H

  • Molecular Weight 185.95

  •  Beilstein Registry Number 1739079

  •  EC Number 200-590-1

  •  MDL number MFCD00002685

  •  PubChem Substance ID 329747113

Purchase

Properties

Related Categories Acids, Acids & Bases, Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography,
InChI Key   JDNTWHVOXJZDSN-UHFFFAOYSA-N
grade   puriss. p.a.
assay   ≥99.5% (T)
impurities   ≤0.1% water
ign. residue   ≤0.02% (as SO4)
mp   77-79 °C(lit.)
  80-84 °C
anion traces   iodide (I-): ≤0.01%
cation traces   Cd: ≤1 mg/kg
  Cu: ≤1 mg/kg
  Fe: ≤1 mg/kg
  Pb: ≤1 mg/kg
  Zn: ≤1 mg/kg
storage temp.   2-8°C

Description

Application

Reagent for the modification of cysteine residues in proteins.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2923 6.1(8) / PGI
WGK Germany 
1
RTECS 
AI3500000

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?