EMAIL THIS PAGE TO A FRIEND
00660 Sigma-Aldrich

2,2-Dimethoxypropane

purum, ≥96.0% (GC)

Synonym: Acetone dimethyl acetal

  • CAS Number 77-76-9

  • Linear Formula (CH3)2C(OCH3)2

  • Molecular Weight 104.15

  •  Beilstein Registry Number 635678

  •  EC Number 201-056-0

  •  MDL number MFCD00008479

  •  PubChem Substance ID 329747114

Purchase

Properties

Related Categories Chemical Synthesis, Other Protecting and Derivatizing Reagents, Protecting and Derivatizing Reagents, Protection and Derivatization, Synthetic Reagents More...
vapor density   3.59 (vs air)
vapor pressure   60 mmHg ( 15.8 °C)
InChI Key   HEWZVZIVELJPQZ-UHFFFAOYSA-N
grade   purum
assay   ≥96.0% (GC)
expl. lim.   31 %, 58 °F
  6 %, 27 °F
refractive index   n20/D 1.378(lit.)
  n20/D 1.378
bp   83 °C(lit.)
density   0.847 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993BE 3 / PGII
WGK Germany 
2
Flash Point(F) 
14 °F
Flash Point(C) 
-10 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

D136808 2,2-Dimethoxypropane, reagent grade, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?