• USA Home
  • 00712 - Acetonitrile solution

EMAIL THIS PAGE TO A FRIEND
00712 Sigma-Aldrich

Acetonitrile solution

~20% in H2O, for protein sequence analysis

  • CAS Number 75-05-8

  • Linear Formula CH3CN

  • Molecular Weight 41.05

  •  Beilstein Registry Number 4124158

  •  MDL number MFCD00001878

  •  PubChem Substance ID 329747123

  •  eCl@ss 39031501

Purchase

Properties

Related Categories Molecular Biology, Protein Sequencing, Protein Structural Analysis, Proteomics, Reagents for Protein Sequencing More...
grade   for protein sequence analysis
InChI Key   WEVYAHXRMPXWCK-UHFFFAOYSA-N
concentration   ~20% in H2O
density   0.97 g/mL at 20 °C
  0.782 g/mL at 25 °C
suitability   in accordance for sequence analysis test
storage temp.   2-8°C

Description

Packaging

Special packaging for direct use in common sequencers.

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1648 3 / PGII
WGK Germany 
2
Flash Point(F) 
42.8 °F
Flash Point(C) 
6 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?