EMAIL THIS PAGE TO A FRIEND
00800 Sigma-Aldrich

Acetophenone

≥98.0% (GC)

Synonym: Methyl phenyl ketone

  • CAS Number 98-86-2

  • Linear Formula CH3COC6H5

  • Molecular Weight 120.15

  •  Beilstein Registry Number 605842

  •  EC Number 202-708-7

  •  MDL number MFCD00008724

  •  PubChem Substance ID 329747143

  •  eCl@ss 39023804

Purchase

Properties

vapor density   4.1 (vs air)
vapor pressure   0.45 mmHg ( 25 °C)
  1 mmHg ( 15 °C)
InChI Key   KWOLFJPFCHCOCG-UHFFFAOYSA-N
assay   ≥98.0% (GC)
autoignition temp.   1058 °F
refractive index   n20/D 1.534(lit.)
  n20/D 1.534
bp   202 °C(lit.)
mp   19-20 °C(lit.)
density   1.03 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3334
WGK Germany 
1
RTECS 
AM5250000
Flash Point(F) 
168.8 °F
Flash Point(C) 
76 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?