• USA Home
  • 00877 - (S)-(−)-2-Acetoxypropionyl chloride

EMAIL THIS PAGE TO A FRIEND
00877 Sigma-Aldrich

(S)-(−)-2-Acetoxypropionyl chloride

for chiral derivatization, ≥99.0%

Synonym: O-Acetyl-L-lactyl chloride, AP-Cl

Purchase

Properties

Related Categories Alcohols, Analytical Reagents, Analytical/Chromatography, Chiral Derivatization Reagents, Chiral GC Derivatization Reagents,
grade   for chiral derivatization
InChI Key   ALHZEIINTQJLOT-VKHMYHEASA-N
assay   ≥99.0% (AT)
  ≥99.0%
optical activity   [α]20/D −33.0±2°, c = 4% in chloroform
optical purity   enantiomeric ratio: ≥97:3 (GC)
refractive index   n20/D 1.423(lit.)
  n20/D 1.423
bp   50 °C/5 mmHg(lit.)
density   1.189 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

Chiral derivatizing agent for the det. of enantiomeric purity of alcohols by GC or NMR

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3265 8 / PGII
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?