• USA Home
  • 00880 - (S)-(−)-2-Acetoxy-1,1,2-triphenylethanol

EMAIL THIS PAGE TO A FRIEND
00880 Sigma-Aldrich

(S)-(−)-2-Acetoxy-1,1,2-triphenylethanol

puriss., ≥99.0% (sum of enantiomers, HPLC)

Synonym: (S)-(−)-1,1,2-Triphenyl-1,2-ethanediol 2-acetate

  • CAS Number 95061-51-1

  • Empirical Formula (Hill Notation) C22H20O3

  • Molecular Weight 332.39

  •  Beilstein Registry Number 4297986

Purchase

Properties

Related Categories Alcohols, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More...
grade   puriss.
assay   ≥99.0% (sum of enantiomers, HPLC)
optical activity   [α]20/D −216±4°, c = 1% in pyridine
mp   ~243 °C (dec.)

Description

Other Notes

Chiral acetate employed in asymmetric aldol condensations

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?