EMAIL THIS PAGE TO A FRIEND
00926 Sigma-Aldrich

Polystyrene

standard, for GPC, 400 certified according to DIN

Synonym: PS

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Application CRMs, Certified Reference Materials (CRMs),
grade   for GPC
  standard
CofA   exact values can be taken from the accompanying certificate of analysis
mol wt   Mn ~370
  Mp ~381
  Mw ~436
Mw/Mn   ~1.18

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?