EMAIL THIS PAGE TO A FRIEND
00939 Sigma-Aldrich

4-Acetylbiphenyl

purum, ≥95.0% (HPLC)

Synonym: 4-Biphenyl methyl ketone, 4-Phenylacetophenone

  • CAS Number 92-91-1

  • Linear Formula C6H5C6H4COCH3

  • Molecular Weight 196.24

  •  Beilstein Registry Number 1101615

  •  EC Number 202-202-6

  •  MDL number MFCD00008749

  •  PubChem Substance ID 329747170

Purchase

Properties

Related Categories Building Blocks, C13 to C14, Carbonyl Compounds, Chemical Synthesis, Ketones,
InChI Key   QCZZSANNLWPGEA-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (HPLC)
mp   116-118 °C(lit.)
  118-122 °C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
AM9662502

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

122726 4-Acetylbiphenyl, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?