EMAIL THIS PAGE TO A FRIEND
00948 Sigma-Aldrich

Polystyrene

standard, for GPC, 70,000 certified according to DIN

Synonym: PS

Purchase

Properties

grade   for GPC
  standard
CofA   exact values can be taken from the accompanying certificate of analysis
mol wt   Mn ~72000
  Mp ~76000
  Mw ~74500
Mw/Mn   ~1.03

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?