EMAIL THIS PAGE TO A FRIEND
01410 Sigma-Aldrich

1-Acetylpiperazine

purum, ≥98.0% (GC)

  • CAS Number 13889-98-0

  • Empirical Formula (Hill Notation) C6H12N2O

  • Molecular Weight 128.17

  •  Beilstein Registry Number 112220

  •  EC Number 237-659-0

  •  MDL number MFCD00058676

Purchase

Properties

Related Categories Building Blocks, C4 to C8, Chemical Synthesis, Heterocyclic Building Blocks, Piperazines More...
InChI Key   PKDPUENCROCRCH-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
mp   31-34 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?