• USA Home
  • 01707 - Acrylamide:N,N′-Methylenebisacrylamide 19:1

EMAIL THIS PAGE TO A FRIEND
01707 Sigma-Aldrich

Acrylamide:N,N′-Methylenebisacrylamide 19:1

for molecular biology

Purchase

Properties

Related Categories Gel Casting Reagents, Isoelectric Focusing and Two Dimensional Electrophoresis, Molecular Biology, Protein Electrophoresis, Proteomics More...
grade   for molecular biology
InChI Key   OCQZXMCGTAWGEQ-UHFFFAOYSA-N
impurities   DNases, none detected
  RNases, none detected
  phosphatases, none detected
  proteases, none detected
conductivity   <20 μS/cm at 20 °C (50 mg/ml water)
solubility   H2O: 50 mg/mL at 20 °C clear, colorless
anion traces   chloride (Cl-): ≤50 mg/kg
  sulfate (SO42-): ≤50 mg/kg
cation traces   Ca: ≤10 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  K: ≤50 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Na: ≤50 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Zn: ≤5 mg/kg
suitability   in accordance for electrophoresis test
storage temp.   2-8°C

Description

Other Notes

DNA sequencing

Price and Availability

Safety & Documentation

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?