EMAIL THIS PAGE TO A FRIEND
01731 Sigma-Aldrich

Acrylamido buffer

≥98.0% (T), pKa 7.4

Purchase

Properties

Related Categories Acrylamido Buffers for IPGs, Isoelectric Focusing and Two Dimensional Electrophoresis, Molecular Biology, Protein Electrophoresis, Proteomics More...
assay   ≥98.0% (T)
shelf life   limited shelf life
pKa    7.4
mp   60-65 °C
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?