EMAIL THIS PAGE TO A FRIEND
01750 Sigma-Aldrich

Ethyl acrylate

purum, ≥99.0% (GC)

Synonym: Acrylic acid ethyl ester

  • CAS Number 140-88-5

  • Linear Formula CH2=CHCOOC2H5

  • Molecular Weight 100.12

  •  Beilstein Registry Number 773866

  •  EC Number 205-438-8

  •  MDL number MFCD00009188

  •  PubChem Substance ID 329747354

Purchase

Properties

Related Categories Building Blocks, C2 to C5, Carbonyl Compounds, Chemical Synthesis, Esters,
vapor density   3.5 (vs air)
vapor pressure   31 mmHg ( 20 °C)
InChI Key   JIGUQPWFLRLWPJ-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (GC)
autoignition temp.   721 °F
contains   ~0.002% hydroquinone monomethyl ether as stabilizer
expl. lim.   12.1 %
refractive index   n20/D 1.406(lit.)
  n20/D 1.406
bp   99 °C(lit.)
mp   −71 °C(lit.)
density   0.921 g/mL at 20 °C
  0.918 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Precautionary statements 
RIDADR 
UN 1917 3 / PGII
WGK Germany 
2
RTECS 
AT0700000
Flash Point(F) 
46.4 °F
Flash Point(C) 
8 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?