EMAIL THIS PAGE TO A FRIEND
01770 Sigma-Aldrich

Butyl acrylate

purum, ≥99.0% (GC)

  • CAS Number 141-32-2

  • Linear Formula CH2=CHCOO(CH2)3CH3

  • Molecular Weight 128.17

  •  Beilstein Registry Number 1749970

  •  EC Number 205-480-7

  •  MDL number MFCD00009446

  •  PubChem Substance ID 329747357

Purchase

Properties

Related Categories Building Blocks, C6 to C7, Carbonyl Compounds, Chemical Synthesis, Esters,
vapor density   >1 (vs air)
vapor pressure   3.3 mmHg ( 20 °C)
InChI Key   CQEYYJKEWSMYFG-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (GC)
autoignition temp.   559 °F
contains   ~0.0015% hydroquinone monomethyl ether as stabilizer
expl. lim.   9.9 %
refractive index   n20/D 1.418(lit.)
  n20/D 1.419
bp   145 °C(lit.)
density   0.899 g/mL at 20 °C
  0.894 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Precautionary statements 
RIDADR 
UN 2348 3 / PGIII
WGK Germany 
1
RTECS 
UD3150000
Flash Point(F) 
96.8 °F
Flash Point(C) 
36 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

234923 Butyl acrylate, ≥99%, contains 10-60 ppm monomethyl ether hydroquinone as inhibitor

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?