EMAIL THIS PAGE TO A FRIEND
01780 Sigma-Aldrich

Acryloyl chloride

purum, ≥96.0% (HPLC)

Synonym: 2-Propenoyl chloride

  • CAS Number 814-68-6

  • Linear Formula CH2=CHCOCl

  • Molecular Weight 90.51

  •  Beilstein Registry Number 635744

  •  EC Number 212-399-0

  •  MDL number MFCD00000731

  •  PubChem Substance ID 24845195

Purchase

Properties

Related Categories Acid Halides, Building Blocks, Carbonyl Compounds, Chemical Synthesis, Organic Building Blocks More...
vapor density   >1 (vs air)
vapor pressure   1.93 psi ( 20 °C)
InChI Key   HFBMWMNUJJDEQZ-UHFFFAOYSA-N
grade   purum
assay   ≥96.0% (HPLC)
contains   ~0.05% phenothiazine as stabilizer
refractive index   n20/D 1.435(lit.)
  n20/D 1.435
bp   72-76 °C(lit.)
density   1.119 g/mL at 20 °C
  1.114 g/mL at 25 °C(lit.)
shipped in   wet ice
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3383 6.1/PG 1
WGK Germany 
3
RTECS 
AT7350000
Flash Point(F) 
57.2 °F
Flash Point(C) 
14 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

A24109 Acryloyl chloride, ≥97%, contains ~400 ppm phenothiazine as stabilizer

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?