EMAIL THIS PAGE TO A FRIEND
01827 Sigma-Aldrich

1-Adamantaneethanol

purum, ≥98.0% (GC)

Synonym: 2-(1-Adamantyl)ethanol

  • CAS Number 6240-11-5

  • Empirical Formula (Hill Notation) C12H20O

  • Molecular Weight 180.29

  •  Beilstein Registry Number 1855382

  •  EC Number 228-353-8

  •  MDL number MFCD00074756

Purchase

Properties

InChI Key   ZBIDZPHRNBZTLT-BKUVIOGVSA-N
grade   purum
assay   ≥98.0% (GC)
mp   66-69 °C(lit.)
  70-74 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

188115 1-Adamantaneethanol, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?