• USA Home
  • 02135 - Zinc bromide solution

EMAIL THIS PAGE TO A FRIEND
02135 Sigma-Aldrich

Zinc bromide solution

optical grade for shielded windows of hot cells, D20 2.5

Purchase

Properties

Related Categories Essential Chemicals, Materials Science, Metal and Ceramic Science, Research Essentials, Salts,
grade   optical grade for shielded windows of hot cells
InChI Key   VNDYJBBGRKZCSX-UHFFFAOYSA-L
quality   D20 2.5
storage temp.   2-8°C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3264 8 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?