EMAIL THIS PAGE TO A FRIEND

02151 Sigma-Aldrich

Sodium bromate

≥99%

Purchase

Properties

Related Categories Chemical Synthesis, Essential Chemicals, Inorganic Salts, Research Essentials, Salts,
assay   ≥99%
impurities   ≤0.005% heavy metals (as Pb)
density   3.339 g/mL at 25 °C(lit.)
anion traces   bromide (Br-): ≤500 mg/kg
  sulfate (SO42-): ≤500 mg/kg
cation traces   Fe: ≤50 mg/kg

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.




Ultra-high Purity Salts

Customers Also Viewed

analytical standard

Chromium(III) chloride hexahydrate

purum p.a., ≥98.0% (RT)

Ferroin indicator solution

redox indicator, Reag. Ph. Eur., E0 in sulfuric acid 1 mol/l = +1,06 volt

Sodium bromate

puriss. p.a., ≥99.5% (RT)

Sodium bromate

≥99%

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Hazard Codes (Europe) 
Risk Statements (Europe) 
Safety Statements (Europe) 
17-26-36
RIDADR 
UN 1494 5.1 / PGII
WGK Germany 
3
RTECS 
EF8750000

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Self-oscillating gel composed of thermosensitive polymer exhibiting higher LCST. Hidaka M and Yoshida R J. Control. Release 150(2), 171-6, (2011)

Loading...


Chemical and optical control of peristaltic actuator based on self-oscillating porous gel. Shinohara S, Seki T, Sakai T, et al. Chem. Commun. (Camb.) (39), 4735-7, (2008)

Loading...


[A case of sodium bromate intoxication with cerebral lesion]. Matsuyama Z, Katayama S, and Nakamura S Rinsho. Shinkeigaku 33(5), 535-40, (1993)

Loading...


[Acute renal failure, neuropathy and deafness due to sodium bromate intoxication: report of one case]. Yei CC, Fuh JJ, Su SY, et al. J. Formos. Med. Assoc. 92 Suppl 4, S262-6, (1993)

Loading...


Intermolecular disulfide cross-linked structural change induced by permanent wave treatment of human hair with thioglycolic acid. Suzuta K, Ogawa S, Takeda Y, et al. J. Cosmet. Sci. 63(3), 177-96, (2012)

Loading...


Acute kidney injury due to sodium bromate intoxication: a report of two cases. Ryu DH, Jang KA, Kim SM, et al. Korean J. Intern. Med. 26(4), 463-5, (2011)

Loading...


Toxicology studies of sodium bromate (CAS No. 7789-38-0) in genetically modified (FVB Tg.AC Hemizygous) mice (dermal and drinking water studies) and carcinogenicity studies of sodium bromate in genetically modified [B6.129-Trp53tm1Brd (N5) haploinsufficient] mice (drinking water studies). Natl. Toxicol. Program Genet. Modif. Model Rep. (6), 1-169, (2007)

Loading...


Two children with bromate intoxication due to ingestion of the second preparation for permanent hair waving. Watanabe T, Abe T, Satoh M, et al. Acta Paediatr. Jpn. 34(6), 601-5, (1992)

Loading...


Propagation of photosensitive chemical waves on the circular routes. Kitahata H, Yamada A, Nakata S, et al. J. Phys. Chem. A 109(22), 4973-8, (2005)

Loading...


Chiral symmetry breaking and complete chiral purity by thermodynamic-kinetic feedback near equilibrium: implications for the origin of biochirality. Viedma C Astrobiology 7(2), 312-9, (2007)

Loading...


Effect of gravity field on the nonequilibrium/nonlinear chemical oscillation reactions. Fujieda S, Mori Y, Nakazawa A, et al. Adv. Space Res. 28(4), 537-43, (2001)

Loading...


Phase III interlaboratory study of FETAX. Part 3. FETAX validation using 12 compounds with and without an exogenous metabolic activation system. Bantle JA, Finch RA, Fort DJ, et al. J. Appl. Toxicol. 19(6), 447-72, (1999)

Loading...


Salt effects on caffeine solubility, distribution, and self-association. Al-Maaieh A and Flanagan DR J. Pharm. Sci. 91(4), 1000-8, (2002)

Loading...


Effects of sodium bromate on ionic concentrations and osmolalities of the cochlear fluids in guinea pigs. Muratsuka Y, Ueda H, and Konishi T Hear. Res. 39(3), 241-9, (1989)

Loading...


Ultramicro-analysis by use of light-scanning photoacoustic densitometry for electrophoresed protein in human hair. Fukami T, Uchiyama K, Yoshimura Y, et al. Anal. Biochem. 238(1), 60-4, (1996)

Loading...


Pattern formation in the Belousov-Zhabotinsky-PAMAM dendrimer system. Roncaglia DI, Carballido-Landeira J, and Muñuzuri AP Phys. Chem. Chem. Phys. 13(16), 7426-32, (2011)

Loading...


A possible role for cell proliferation in potassium bromate (KBrO3) carcinogenesis. Umemura T, Sai K, Takagi A, et al. J. Cancer Res. Clin. Oncol. 119(8), 463-9, (1993)

Loading...


Nonequilibrium / nonlinear chemical oscillation in the virtual absence of gravity. Fujieda S, Mogami Y, Moriyasu K, et al. Adv. Space Res. 23(12), 2057-63, (1999)

Loading...


Ternary phase diagram for the Belousov-Zhabotinsky reaction-induced mechanical oscillation of intelligent PNIPAM colloids. Shen J, Pullela S, Marquez M, et al. J. Phys. Chem. A 111(48), 12081-5, (2007)

Loading...


Release of two peripheral proteins from chloroplast thylakoid membranes in the presence of a Hofmeister series of chaotropic anions. Hincha DK Arch. Biochem. Biophys. 358(2), 385-90, (1998)

Loading...


Vibrational investigation of the stretching region of bromate ion in solution. Alves WA and Faria RB Spectrochim. Acta. A. Mol. Biomol. Spectrosc. 58(7), 1395-9, (2002)

Loading...


Immunotoxicity of sodium bromate in female B6C3F1 mice: a 28-day drinking water study. Guo TL, McCay JA, Karrow NA, et al. Drug Chem. Toxicol. 24(2), 129-49, (2001)

Loading...


An elderly patient with severe acute renal failure due to sodium bromate intoxication. Uchida HA, Sugiyama H, Kanehisa S, et al. Intern. Med. 45(3), 151-4, (2006)

Loading...


Acute renal failure and hearing loss due to sodium bromate poisoning: a case report and review of the literature. Sashiyama H, Irie Y, Ohtake Y, et al. Clin. Nephrol. 58(6), 455-7, (2002)

Loading...


Bromate intoxication with polyneuropathy. Wang V, Lin KP, Tsai CP, et al. J. Neurol. Neurosurg. Psychiatr. 58(4), 516-7, (1995)

Loading...


Merck 14,8593

Fieser 1,1055

Corp MSDS 1 (2), 3134:A / FT-IR 2 (3), 4763:D / FT-IR 1 (2), 1254:C / RegBook 1 (3), 3461:C / Sax 6, 2415

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?