EMAIL THIS PAGE TO A FRIEND
02190 Sigma-Aldrich

Adipoyl chloride

purum, ≥99.0% (T)

Synonym: Adipyl chloride

  • CAS Number 111-50-2

  • Linear Formula ClCO(CH2)4COCl

  • Molecular Weight 183.03

  •  Beilstein Registry Number 507709

  •  EC Number 203-876-4

  •  MDL number MFCD00000759

  •  PubChem Substance ID 329747400

Purchase

Properties

Related Categories Acid Halides, Building Blocks, Carbonyl Compounds, Chemical Synthesis, Organic Building Blocks More...
InChI Key   PWAXUOGZOSVGBO-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (T)
refractive index   n20/D 1.471(lit.)
  n20/D 1.472
bp   105-107 °C/2 mmHg(lit.)
density   1.259 g/mL at 25 °C(lit.)

Description

Caution

may discolor to yellow-brown on storage

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3265 8 / PGII
WGK Germany 
3
Flash Point(F) 
233.6 °F
Flash Point(C) 
112 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?