• USA Home
  • 02352 - Esculin sesquihydrate

EMAIL THIS PAGE TO A FRIEND
02352 Sigma-Aldrich

Esculin sesquihydrate

tested according to DAB

Synonym: 6,7-Dihydroxycoumarin 6-glucoside, Aesculin, Aesculinum, Esculetin-6-β-D-glucopyranoside

  • CAS Number 66778-17-4

  • Empirical Formula (Hill Notation) C15H16O9 · 1.5H2O

  • Molecular Weight 367.31

  •  Beilstein Registry Number 95387

  •  MDL number MFCD00149492

  •  PubChem Substance ID 24845257

Purchase

Properties

Related Categories Carbohydrate Chemistry, Carbohydrate Derivatives, Chemical Biology, Chemical Synthesis
InChI Key   CQYPGSKIFJFVDQ-QWFKVUSTSA-N
pharmacopeia   testing conforms to Pharmacopeia

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?