• USA Home
  • 02355 - Poly(methacrylic acid sodium salt)

EMAIL THIS PAGE TO A FRIEND
02355 Sigma-Aldrich

Poly(methacrylic acid sodium salt)

analytical standard, for GPC, 7,750

Synonym: Poly(sodium methacrylate)

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Poly(methylacrylic acid sodium salt), Polymer Standards,
grade   analytical standard
  for GPC
CofA   exact values can be taken from the accompanying certificate of analysis
shelf life   limited shelf life, expiry date on the label
mol wt   Mn ~7220
  Mp ~7830
  Mw ~7750
Mw/Mn   ~1.07
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?