• USA Home
  • 02357 - Poly(methacrylic acid sodium salt)

EMAIL THIS PAGE TO A FRIEND
02357 Sigma-Aldrich

Poly(methacrylic acid sodium salt)

analytical standard, for GPC, 31,100

Synonym: Poly(sodium methacrylate)

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Poly(methylacrylic acid sodium salt), Polymer Standards,
grade   analytical standard
  for GPC
CofA   exact values can be taken from the accompanying certificate of analysis
mol wt   Mn ~30400
  Mp ~31500
  Mw ~31100
Mw/Mn   ~1.02

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?