• USA Home
  • 02397 - Sodium standard for AAS

EMAIL THIS PAGE TO A FRIEND
02397 Sigma-Aldrich

Sodium standard for AAS

analytical standard, 10.00 g/L Na in nitric acid

Synonym: Sodium nitrate solution

Purchase

Properties

Related Categories AAS, Additional Standards, Analytical Standards, Analytical/Chromatography, Elemental & Metal Standards,
grade   analytical standard
description   ready-to-use
InChI Key   KEAYESYHFKHZAL-UHFFFAOYSA-N
shelf life   limited shelf life, expiry date on the label
concentration   10.00 g/L Na in nitric acid
density   1.04 g/mL at 20 °C

Description

Preparation Note

prepared with NaNO3 and HNO3 (~0.5 M)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3264 8 / PGIII
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?