• USA Home
  • 02420 - Ethanesulfonyl chloride

EMAIL THIS PAGE TO A FRIEND
02420 Sigma-Aldrich

Ethanesulfonyl chloride

purum, ≥98.0% (AT)

  • CAS Number 594-44-5

  • Linear Formula CH3CH2SO2Cl

  • Molecular Weight 128.58

  •  Beilstein Registry Number 773865

  •  EC Number 209-842-5

  •  MDL number MFCD00007460

  •  PubChem Substance ID 24845278

Purchase

Properties

Related Categories Chemical Synthesis, Other Protecting and Derivatizing Reagents, Protecting and Derivatizing Reagents, Protection and Derivatization, Synthetic Reagents More...
InChI Key   FRYHCSODNHYDPU-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (AT)
refractive index   n20/D 1.452(lit.)
  n20/D 1.453
bp   177 °C(lit.)
density   1.357 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2927 8(6.1) / PGII
WGK Germany 
3
RTECS 
KI8050000
Flash Point(F) 
181.4 °F
Flash Point(C) 
83 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

471542 Ethanesulfonyl chloride, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?