EMAIL THIS PAGE TO A FRIEND
02424 Sigma-Aldrich

Ethanesulfonic acid

purum, ≥99.0% (T)

  • CAS Number 594-45-6

  • Linear Formula CH3CH2SO3H

  • Molecular Weight 110.13

  •  Beilstein Registry Number 1743071

  •  EC Number 209-843-0

  •  MDL number MFCD00007529

  •  PubChem Substance ID 24845280

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfonic/Sulfinic Acids, Sulfur Compounds More...
InChI Key   CCIVGXIOQKPBKL-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (T)
refractive index   n20/D 1.434(lit.)
  n20/D 1.434
bp   123 °C/0.01 mmHg(lit.)
mp   −17 °C(lit.)
density   1.35 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2586PSN1 8 / PGIII
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

186260 Ethanesulfonic acid, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?