• USA Home
  • 02452 - N-Methyl-L-phenylalanine hydrochloride

EMAIL THIS PAGE TO A FRIEND
02452 Sigma-Aldrich

N-Methyl-L-phenylalanine hydrochloride

≥98.0% (TLC)

  • CAS Number 66866-67-9

  • Empirical Formula (Hill Notation) C10H13NO2 · HCl

  • Molecular Weight 215.68

  •  Beilstein Registry Number 4018032

  •  MDL number MFCD00754494

  •  PubChem Substance ID 24845284

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, N-Methyl Amino Acids, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   DFQSBSADGQZNFW-FVGYRXGTSA-N
assay   ≥98.0% (TLC)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?