• USA Home
  • 02457 - Calcium standard for AAS

EMAIL THIS PAGE TO A FRIEND
02457 Sigma-Aldrich

Calcium standard for AAS

analytical standard, ready-to-use, traceable to BAM, in nitric acid

Synonym: Calcium nitrate solution

Purchase

Properties

Related Categories AAS, Additional Standards, Analytical Standards, Analytical/Chromatography, Elemental & Metal Standards,
grade   analytical standard
InChI Key   ZCCIPPOKBCJFDN-UHFFFAOYSA-N
shelf life   limited shelf life, expiry date on the label
concentration   in nitric acid
  10.00 g/L Ca+2
density   1.04 g/mL at 20 °C

Description

Preparation Note

prepared with Ca(NO3)2.4H2O and HNO3 (~0.5 M)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3264 8 / PGIII
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?