EMAIL THIS PAGE TO A FRIEND
02581 Sigma-Aldrich

Methanol solution

for protein sequence analysis, ~50% in H2O

  • CAS Number 67-56-1

  • Empirical Formula (Hill Notation) CH4O

  • Molecular Weight 32.04

  •  Beilstein Registry Number 3662861

  •  MDL number MFCD00004595

Purchase

Properties

Related Categories Molecular Biology, Protein Sequencing, Protein Structural Analysis, Proteomics, Reagents for Protein Sequencing More...
grade   for protein sequence analysis
InChI Key   OKKJLVBELUTLKV-UHFFFAOYSA-N
concentration   ~50% in H2O
suitability   in accordance for sequence analysis test

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability


Detect Your Unseen Protein
Safety & Documentation

Safety Information

RIDADR 
UN 1230 3/PG 2
WGK Germany 
1
Flash Point(F) 
52 °F
Flash Point(C) 
11 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?