• USA Home
  • 02593 - Polyamide with Fluorescence

EMAIL THIS PAGE TO A FRIEND
02593 Sigma-Aldrich

Polyamide with Fluorescence

for thin layer chromatography, 6 DF, with fluorescent indicator 254 nm

Purchase

Properties

Related Categories Adsorbents for TLC, Analytical/Chromatography, LPLC/TLC/Paper Chromatography, Polyamide 6 (Nylon 6) TLC Adsorbents, Thin Layer Chromatography (TLC) More...
grade   for thin layer chromatography
quality   with fluorescent indicator 254 nm
bulk density   0.25 g/mL

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?