• USA Home
  • 02612 - (Ethoxycarbonylmethyl)triphenylphosphonium bromide

EMAIL THIS PAGE TO A FRIEND
02612 Sigma-Aldrich

(Ethoxycarbonylmethyl)triphenylphosphonium bromide

purum, 97% (AT)

Synonym: (Carboxymethyl)triphenylphosphonium bromide ethyl ester, NSC 60450

  • CAS Number 1530-45-6

  • Linear Formula CH3CH2OCOCH2P(C6H5)3Br

  • Molecular Weight 429.29

  •  Beilstein Registry Number 3581273

  •  EC Number 216-230-1

  •  MDL number MFCD00011835

Purchase

Properties

InChI Key   VJVZPTPOYCJFNI-UHFFFAOYSA-M
grade   purum
assay   97% (AT)
mp   145-150 °C (dec.)(lit.)
  145-150 °C (dec.)
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Reactant for:
• Wittig olefination reactions for synthesis of tetramic acid derivatives
• Synthesis of precursors for Bergman cyclization
• Preparation of PPAR agonists derived from thalidomide-related LXR antagonists
• Synthesis of benzoxaboroles with antimalarial activity
• Michael acceptor based cysteine protease inhibitors
• Mechanistic studies of the IspH reductase protein

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
TA2299500

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

412252 (Ethoxycarbonylmethyl)triphenylphosphonium bromide, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?