• USA Home
  • 02675 - N-Methyl-L-isoleucine

EMAIL THIS PAGE TO A FRIEND
02675 Sigma-Aldrich

N-Methyl-L-isoleucine

≥98.0%

  • CAS Number 4125-98-8

  • Empirical Formula (Hill Notation) C7H15NO2

  • Molecular Weight 145.20

  •  Beilstein Registry Number 1722062

  •  MDL number MFCD00037755

  •  PubChem Substance ID 329747486

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, N-Methyl Amino Acids, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   KSPIYJQBLVDRRI-WDSKDSINSA-N
assay   ≥98.0%

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?